Learn More
ACROBiosystemsSupplier Diversity Partner Recombinant Protein;Biotinylated Human LILRB2 / CD85d / ILT4 Protein, Fc Tag, Avi Tag (Avitag™);HEK293;25UG;LI2,LILRB2,ILT4,LIR2,MIR10,CD85d,LIR-2,ILT-4,MIR-10  DFS Item
Manufacturer:  ACROBiosystemsSupplier Diversity Partner LI2H82F525UG
Recombinant Protein;Biotinylated Human LILRB2 / CD85d / ILT4 Protein, Fc Tag, Avi Tag (Avitag™);25UG;Biotinylated Human LILRB2, Fc Tag (LI2-H82F5) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Val 461 (Accession # AAH36827.1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 76.2 kDa. The protein migrates as 36 kDa and 100-116 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human ANGPTL7, His Tag (Cat. No. AN7-H52H5) at 5 μg/mL (100 μL/well) can bind Biotinylated Human LILRB2, Fc Tag (Cat. No. LI2-H82F5) with a linear range of 0.156-2.5 μg/mL (QC tested).
Catalog No. 50-165-8517