Learn More
ACROBiosystemsSupplier Diversity Partner Recombinant Protein;Vibrio cholerae serotype O1 CTXB Protein;HEK293;50UG;CTB,CTXB,TOXB,Cholera enterotoxin  DFS Item
Manufacturer:  ACROBiosystemsSupplier Diversity Partner CTBV52E850UG
Recombinant Protein;Vibrio cholerae serotype O1 CTXB Protein;50UG;Vibrio cholerae serotype O1 CTXB, His Tag (CTB-V52E8) is expressed from human 293 cells (HEK293). It contains AA Thr 22 - Asn 124 (Accession # P01556).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 13.5 kDa. The protein migrates as 18-22 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-165-8558