Learn More
ACROBiosystems Recombinant Protein;Human Lumican Protein;HEK293;100UG;LUM,LUM,Lumican,LDC,SLRR2D,KSPG lumican
Supplier:  ACROBiosystems LUMH5227100UG
Encompass_Preferred
Recombinant Protein;Human Lumican Protein;100UG;Human Lumican, His Tag (LUM-H5227) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Asn 338 (Accession # NP_002336).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 37.5 kDa. The protein migrates as 50-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8569
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.