Learn More
ACROBiosystems Recombinant Protein;Human FABP3 / H-FABP Protein;E.coli;100UG;FA3,FABP3,H-FABP,FABP11,MDGI,M-FABP,FABP-3,hFABP,O-FABP
Supplier: ACROBiosystems FA3H5128100UG
Recombinant Protein;Human FABP3 / H-FABP Protein;100UG;Human FABP3, His Tag (FA3-H5128) is expressed from E.coli cells. It contains AA Met 1 - Ala 133 (Accession # NP_004093).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 15.7 kDa. The protein migrates as 16 kDa under reducing (R) condition (SDS-PAGE).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.