Learn More
ACROBiosystems Recombinant Protein;Human DLL4 Protein;HEK293;100UG;DL4,DLL4,Delta4,DLL-4,Delta-4
Supplier:  ACROBiosystems DL4H5227100UG
Encompass_Preferred
Recombinant Protein;Human DLL4 Protein;100UG;Human DLL4, His Tag (DL4-H5227) is expressed from human 293 cells (HEK293). It contains AA Ser 27 - Pro 524 (Accession # AAI06951).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 55.1 kDa. The protein migrates as 65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8590
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.