Learn More
ACROBiosystems Recombinant Protein;Rhesus macaque CD19 (20-292) Protein, Fc Tag;HEK293;100UG;CD9,CD19,CD-19,B4,B-4,CVID3,CVID-3,MGC12802
Supplier:  ACROBiosystems CD9C5251100UG
Encompass_Preferred
Recombinant Protein;Rhesus macaque CD19 (20-292) Protein, Fc Tag;100UG;Rhesus macaque CD19, Fc Tag (CD9-C5251) is expressed from human 293 cells (HEK293). It contains AA Pro 20 - Lys 292 (Accession # F7F486).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 56.7kDa. The protein migrates as 66-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8600