Learn More
ACROBiosystemsSupplier Diversity Partner Recombinant Protein;Recombinant Influenza A [A/Thailand/1(KAN-1)/2004 (H5N1)] Neuraminidase (NA);HEK293;100UG;H54,NA,Neuraminidase,Influenza A,H5N1
Manufacturer:  ACROBiosystemsSupplier Diversity Partner H54V5247100UG
Encompass_Preferred
Recombinant Protein;Recombinant Influenza A [A/Thailand/1(KAN-1)/2004 (H5N1)] Neuraminidase (NA);100UG;Influenza Neuraminidase (NA), His Tag (H54-V5247) is expressed from human 293 cells (HEK293). It contains AA His 36 - Lys 449 (Accession # H8PF47).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 46.1 kDa. The protein migrates as 48 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-165-8602