Learn More
ACROBiosystemsSupplier Diversity Partner Recombinant Protein;Recombinant Influenza A [A/Guinea fowl/Hong Kong/WF10/99(H9N2)] Hemagglutinin (HA);HEK293;100UG;HA2,HA,Hemagglutinin
Manufacturer:  ACROBiosystemsSupplier Diversity Partner HA2V5227100UG
Encompass_Preferred
Recombinant Protein;Recombinant Influenza A [A/Guinea fowl/Hong Kong/WF10/99(H9N2)] Hemagglutinin (HA);100UG;Influenza , His Tag (HA2-V5227) is expressed from human 293 cells (HEK293). It contains AA Asp 19 - Lys 523 (Accession # Q80KD9).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 58.7 kDa. The protein migrates as 60-90 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-165-8606