Learn More
ACROBiosystemsSupplier Diversity Partner Recombinant Protein;Recombinant Influenza A [A/Guinea fowl/Hong Kong/WF10/99 (H9N2)] Hemagglutinin 1(HA1);HEK293;100UG;HA2,HA,Hemagglutinin  DFS Item
Manufacturer:  ACROBiosystemsSupplier Diversity Partner HA2V52H8100UG
Recombinant Protein;Recombinant Influenza A [A/Guinea fowl/Hong Kong/WF10/99 (H9N2)] Hemagglutinin 1(HA1);100UG;Influenza , His Tag (HA2-V52H8) is expressed from human 293 cells (HEK293). It contains AA Asp 19 - Arg 338 (Accession # Q80KD9).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 37.5 kDa. The protein migrates as 50-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8608