Learn More
ACROBiosystems Recombinant Protein;Human FOLR1 Protein, Strep Tag;HEK293;25UG;FO1,FOLR-1,FBP,FOLR,FOLR1
Supplier:  ACROBiosystems FO1H528B25UG
Encompass_Preferred
Recombinant Protein;Human FOLR1 Protein, Strep Tag;25UG;Human FOLR1, Strep Tag (FO1-H528b) is expressed from human 293 cells (HEK293). It contains AA Arg 25 - Met 233 (Accession # NP_057937.1).;This protein carries a twin strep tag at the C-terminus. The protein has a calculated MW of 27.6 kDa. The protein migrates as 33-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8622