Learn More
ACROBiosystems Recombinant Protein;Human VEGF R2 / KDR Protein, Strep Tag;HEK293;25UG;KDR,KDR,CD309,FLK1,VEGFR,VEGFR2,VEGF R
Supplier:  ACROBiosystems KDRH528025UG
Encompass_Preferred
Recombinant Protein;Human VEGF R2 / KDR Protein, Strep Tag;25UG;Human VEGF R2, Strep Tag (KDR-H5280) is expressed from human 293 cells (HEK293). It contains AA Ala 20 - Glu 764 (Accession # AAI31823).;This protein carries a twin strep tag at the C-terminus. The protein has a calculated MW of 86.3 kDa. The protein migrates as 100-110 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized ActiveMax Human VEGF121 (Cat. No. VE1-H4213) at 2 g/mL can bind Human VEGF R2, Strep Tag (Cat. No. KDR-H5280) with a linear range of 0.15-2.5 g/mL (QC tested).
Catalog No. 50-165-8665