Learn More
ACROBiosystemsSupplier Diversity Partner Recombinant Protein;Mouse OX40 / TNFRSF4 / CD134 Protein, Fc Tag;HEK293;100UG;OX0,TNFRSF4,OX40,CD134,OX40L receptor,ACT35,TXGP1L,OX-40
Manufacturer:  ACROBiosystemsSupplier Diversity Partner OX0M5259100UG
Encompass_Preferred
Recombinant Protein;Mouse OX40 / TNFRSF4 / CD134 Protein, Fc Tag;100UG;Mouse OX40, Fc Tag (OX0-M5259) is expressed from human 293 cells (HEK293). It contains AA Val 20 - Pro 211 (Accession # NP_035789).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 47.9 kDa. The protein migrates as 70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse OX40 Ligand, His Tag (Cat. No. TN4-M5241) at 0.5 μg/mL (100 μL/well) can bind Mouse OX40, Fc Tag (Cat. No. OX0-M5259) with a linear range of 10-16 ng/mL (QC tested).
Catalog No. 50-165-8703