Learn More
ACROBiosystems Recombinant Protein;Mouse IL-13 R alpha 1 Protein, Fc Tag;HEK293;200UG;IL1,IL13RA1,CD213A1,IL-13Ra,NR4,RP13-128O4.2,IL13Rα1,Interleukin
Supplier:  ACROBiosystems IL1M5258200UG
Encompass_Preferred
Recombinant Protein;Mouse IL-13 R alpha 1 Protein, Fc Tag;200UG;Mouse IL-13RA1, Fc Tag (IL1-M5258) is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Thr 340 (Accession # NP_598751).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 62.7 kDa. The protein migrates as 67-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8712
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.