Learn More
ACROBiosystems Recombinant Protein;Mouse gp130 / CD130 / IL-6 R beta Protein, Fc Tag;HEK293;100UG;ILT,IL6ST,gp130,CD130,IL-6RB,IL-6R-beta,IL-6R subunit beta,IL-6 receptor subunit beta,CDw130,IL-6ST,IL6RB
Manufacturer:  ACROBiosystems ILTM5252100UG
Encompass_Preferred
Recombinant Protein;Mouse gp130 / CD130 / IL-6 R beta Protein, Fc Tag;100UG;Mouse gp130, Fc Tag (ILT-M5252) is expressed from human 293 cells (HEK293). It contains AA Gln 23 - Glu 617 (Accession # NP_034690).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 93.3 kDa. The protein migrates as 120-130 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8716