Learn More
ACROBiosystemsSupplier Diversity Partner Recombinant Protein;Mouse Dkk-1 Protein;HEK293;50UG;DK1,DKK1,DKK-1,SK  DFS Item
Manufacturer:  ACROBiosystemsSupplier Diversity Partner DK1M52H650UG
Recombinant Protein;Mouse Dkk-1 Protein;50UG;Mouse Dkk-1, His Tag (DK1-M52H6) is expressed from human 293 cells (HEK293). It contains AA Thr 32 - His 272 (Accession # O54908).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 28 kDa. The protein migrates as 41-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8731