Learn More
ACROBiosystemsSupplier Diversity Partner Recombinant Protein;Human FAM171B Protein;HEK293;100UG;FAB,FAM171B,KIAA1946,NPD019
Manufacturer:  ACROBiosystemsSupplier Diversity Partner FABH5220100UG
Encompass_Preferred
Recombinant Protein;Human FAM171B Protein;100UG;Human FAM171B, His Tag (FAB-H5220) is expressed from human 293 cells (HEK293). It contains AA Glu 33 - Thr 355 (Accession # AAH60872).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 37 kDa. The protein migrates as 45-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8763