Learn More
ACROBiosystemsSupplier Diversity Partner Recombinant Protein;Human IL-17 RA / CD217 Protein, Fc Tag;HEK293;200UG;ILA,CD217,CDw217,IL-17RA,CANDF5,IL17R,hIL-17R,IL17RA,Interleukin
Manufacturer:  ACROBiosystemsSupplier Diversity Partner ILAH5257200UG
Encompass_Preferred
Recombinant Protein;Human IL-17 RA / CD217 Protein, Fc Tag;200UG;Human IL-17 RA, Fc Tag (ILA-H5257) is expressed from human 293 cells (HEK293). It contains AA Leu 33 - Trp 320 (Accession # NP_055154.3).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 60.1 kDa. The protein migrates as 71-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL17 /IL17A,Tag Free (Cat. No. ILA-H5118) at 2 μg/mL (100 μL/well ) can bind Human IL-17 RA, Fc Tag (Cat. No. ILA-H5257) with a linear ranger of 0.3-5 ng/mL (QC tested).
Catalog No. 50-165-8773