Learn More
ACROBiosystemsSupplier Diversity Partner Recombinant Protein;Human UCH-L3 / UCHL3 Protein;E.coli;100UG;UC3,UCHL3,UCH-L3,Ubiquitin thioesterase L3  DFS Item
Manufacturer:  ACROBiosystemsSupplier Diversity Partner UC3H5141100UG
Recombinant Protein;Human UCH-L3 / UCHL3 Protein;100UG;Human UCH-L3, His Tag (UC3-H5141) is expressed from E.coli cells. It contains AA Glu 2 - Ala 230 (Accession # AAH18125).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 27 kDa. The protein migrates as 27 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-165-8824