Learn More
ACROBiosystemsSupplier Diversity Partner Recombinant Protein;Human UCH-L1 Protein;E.coli;50UG;UC1,UCHL1,PGP9.5,UCH-L1  DFS Item
Manufacturer:  ACROBiosystemsSupplier Diversity Partner UC1H514050UG
Recombinant Protein;Human UCH-L1 Protein;50UG;Human UCH-L1, His Tag (UC1-H5140) is expressed from E.coli cells. It contains AA Met 1 - Ala 223 (Accession # NP_004172).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 25.7 kDa. The protein migrates as 26 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-165-8826