Learn More
ACROBiosystems Recombinant Protein;Human Niemann-Pick Type C2 / NPC2 Protein;HEK293;100UG;NP2,NPC2,HE1,NPC-2,HE-1
Supplier: ACROBiosystems NP2H52H1100UG
Recombinant Protein;Human Niemann-Pick Type C2 / NPC2 Protein;100UG;Human NPC2, His Tag (NP2-H52H1) is expressed from human 293 cells (HEK293). It contains AA Glu 20 - Leu 151 (Accession # AAH02532).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 16.5 kDa. The protein migrates as 20-24 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.