Learn More
ACROBiosystems Recombinant Protein;Human S100P / S100E Protein, Tag Free;E.coli;100UG;S1P,S100P,S100E,MIG9,S100-P,S100-E
Supplier:  ACROBiosystems S1PH5110100UG
Encompass_Preferred
Recombinant Protein;Human S100P / S100E Protein, Tag Free;100UG;Human S100P, Tag Free (S1P-H5110) is expressed from E.coli cells. It contains AA Met 1 - Lys 95 (Accession # AAH06819).;This protein carries no "tag". The protein has a calculated MW of 10.4 kDa. The protein migrates as 10.4 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-165-8863