Learn More
ACROBiosystemsSupplier Diversity Partner Recombinant Protein;Human SOD2 / Mn-SOD Protein;E.coli;100UG;SO2,SOD2,Mn-SOD
Manufacturer:  ACROBiosystemsSupplier Diversity Partner SO2H5126100UG
Encompass_Preferred
Recombinant Protein;Human SOD2 / Mn-SOD Protein;100UG;Human SOD2, His Tag (SO2-H5126) is expressed from E.coli cells. It contains AA Lys 25 - Lys 222 (Accession # AAH12423).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 23.2 kDa. The protein migrates as 27 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-165-8877