Learn More
ACROBiosystemsSupplier Diversity Partner Recombinant Protein;Human SOD1 / Cu-Zn SOD Protein;E.coli;100UG;SO1,SOD1,ALS,ALS1,HEL-S-44,IPOA,SOD,hSod1,SOD-1,Superoxide dismutase 1
Manufacturer:  ACROBiosystemsSupplier Diversity Partner SO1H5148100UG
Encompass_Preferred
Recombinant Protein;Human SOD1 / Cu-Zn SOD Protein;100UG;Human SOD1, His Tag (SO1-H5148) is expressed from E.coli cells. It contains AA Ala 2 - Gln 154 (Accession # AAH01034).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 16.8 kDa. The protein migrates as 20 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-165-8879