Learn More
ACROBiosystems Recombinant Protein;Human NME1 / NDKA / NM23-H1 Protein;E.coli;100UG;NM1,NME1,NDK A,NDP kinase A,NDPA,GAAD,NM23-H1
Supplier: ACROBiosystems NM1H5147100UG
Recombinant Protein;Human NME1 / NDKA / NM23-H1 Protein;100UG;Human NME1, His Tag (NM1-H5147) is expressed from E.coli cells. It contains AA Ala 2 - Glu 152 (Accession # AAH00293).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 18 kDa. The protein migrates as 21 kDa under reducing (R) condition (SDS-PAGE).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.