Learn More
ACROBiosystemsSupplier Diversity Partner Recombinant Protein;Human IL17 / IL17A Protein, Tag Free ;E.coli;25UG;ILA,IL-17A,Interleukin-17A,CTLA-8,IL-17,IL17A,Interleukin 17A,CTLA8,IL17  DFS Item
Manufacturer:  ACROBiosystemsSupplier Diversity Partner ILAH511825UG
Recombinant Protein;Human IL17 / IL17A Protein, Tag Free ;25UG;Human IL-17A, Tag Free (ILA-H5118) is expressed from E.coli cells. It contains AA Gly 24 - Ala 155 (Accession # Q16552).;This protein carries no "tag". The protein has a calculated MW of 15.3 kDa. the protein migrates as 18 kDa under reducing (R) condition, and 28 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Human IL-17A, Tag Free (Cat. No. ILA-H5118) at 2 μg/mL (100 μL/well) can bind Anti-IL17A MAb, Human IgG1 with a linear range of 0.08-1 ng/mL (Routinely tested).;Immobilized Human IL-17A, Tag Free (Cat. No. ILA-H5118) at 0.5 μg/mL (100 μL/well) can bind Human IL-17 RA, His Tag (Cat. No. ILA-H5222) with a linear range of 8-125 ng/mL (QC tested).;Human IL17/ IL17A Protein, Free Tag(Cat. No. ILA-H5118) stimulates secretion of IL-6 by NIH-3T3 cells. The ED50 for this effect is 1.444-6.941 ng/ml.
Catalog No. 50-165-8904