Learn More
ACROBiosystems Recombinant Protein;Human CD36 / SR-B3 Protein;HEK293;100UG;CD6,CD36,SCARB3,BDPLT10,CHDS7,FAT,GP3B,GP4,GPIV,PASIV,Platelet Glycoprotein 4,glycoprotein IV,gpIV,glycoprotein IIIb,gpIIIb
Supplier:  ACROBiosystems CD6H5221100UG
Encompass_Preferred
Recombinant Protein;Human CD36 / SR-B3 Protein;100UG;Human CD36, His Tag (CD6-H5221) is expressed from human 293 cells (HEK293). It contains AA Gly 30 - Asn 439 (Accession # NP_001001547.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 47.5 kDa. The protein migrates as 66-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8938