Learn More
ACROBiosystems Recombinant Protein;Human BTLA (31-134) Protein, Fc Tag;HEK293;100UG;BTA,BTLA,CD272
Supplier:  ACROBiosystems BTAH5255100UG
Encompass_Preferred
Recombinant Protein;Human BTLA (31-134) Protein, Fc Tag;100UG;Human BTLA, Fc Tag (BTA-H5255) is expressed from human 293 cells (HEK293). It contains AA Lys 31 - Thr 134 (Accession # NP_001078826.1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 38.7 kDa. The protein migrates as 45-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human HVEM, His Tag (Cat.No. HVM-H52E9) at 5 μg/mL (100 μL/well) can bind Human BTLA, Fc Tag (Cat.No. BTA-H5255) with a linear range of 0.08-0.3 μg/mL (QC tested).
Catalog No. 50-165-9004