Learn More
ACROBiosystemsSupplier Diversity Partner Recombinant Protein;Human BMPR-IA / ALK-3 Protein, Fc Tag;HEK293;200UG;AL3,BMPR-1A,BMPR-ⅠA,BMPRⅠA,ALK-3,BMPR1A,BMPRIA,BMPR-IA,10q23del,ACVRLK3,ALK3,CD292,SKR5,ACVRLK-3,ALK-3,SKR-5  DFS Item
Manufacturer:  ACROBiosystemsSupplier Diversity Partner AL3H5259200UG
Recombinant Protein;Human BMPR-IA / ALK-3 Protein, Fc Tag;200UG;Human BMPR-IA, Fc Tag (AL3-H5259) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Arg 152 (Accession # CAA80257).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 42 kDa. The protein migrates as 56 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-9006