Learn More
ACROBiosystemsSupplier Diversity Partner Recombinant Protein;Human BMPR-IA / ALK-3 Protein;HEK293;200UG;AL3,BMPR-1A,BMPR-ⅠA,BMPRⅠA,ALK-3,BMPR1A,BMPRIA,BMPR-IA,10q23del,ACVRLK3,ALK3,CD292,SKR5,ACVRLK-3,ALK-3,SKR-5  DFS Item
Manufacturer:  ACROBiosystemsSupplier Diversity Partner AL3H5228200UG
Recombinant Protein;Human BMPR-IA / ALK-3 Protein;200UG;Human BMPR-IA, His Tag (AL3-H5228) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Arg 152 (Accession # CAA80257).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 15.7 kDa. The protein migrates as 23-34 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-9008