Learn More
ACROBiosystems Recombinant Protein;Human BMP-2 Protein, Tag Free;E.coli;20UG;BM2,BMP2,BMP-2,BMP2A,BMP-2A
Supplier:  ACROBiosystems BM2H411720UG
Encompass_Preferred
Recombinant Protein;Human BMP-2 Protein, Tag Free;20UG;Human BMP-2, Tag Free (BM2-H4117) is expressed from E.coli cells. It contains AA Gln 283 - Arg 396 (Accession # NP_001191).;This protein carries no "tag". The protein has a calculated MW of 13 kDa. the protein migrates as 13 kDa under reducing (R) condition, and 24 kDa under non-reducing (NR) condition (SDS-PAGE).
Catalog No. 50-165-9010
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.