Learn More
ACROBiosystems Recombinant Protein;Human Betacellulin Protein, Fc Tag;HEK293;50UG;BEN,betacellulin,BTC
Supplier:  ACROBiosystems BENH525X50UG
Encompass_Preferred
Recombinant Protein;Human Betacellulin Protein, Fc Tag;50UG;Human Betacellulin, Fc Tag (BEN-H525x) is expressed from human 293 cells (HEK293). It contains AA Asp 32 - Tyr 111 (Accession # AAH11618).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 36.5 kDa. The protein migrates as 45-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-9012