Learn More
ACROBiosystems Recombinant Protein;Human Annexin A5 Protein, Tag Free;E.coli;100UG;AN5,ANXA5,ANXA-5,ANX5,ANX-5,ENX2,ENX-2,PP4,PP-4,annexin V,Annexin A5
Supplier:  ACROBiosystems AN5H5118100UG
Encompass_Preferred
Recombinant Protein;Human Annexin A5 Protein, Tag Free;100UG;Human ANXA5, Tag Free (AN5-H5118) is expressed from E.coli cells. It contains AA Met 1 - Aps 320 (Accession # AAH01429).;This protein carries no "tag". The protein has a calculated MW of 36 kDa. The protein migrates as 35 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-165-9053