Learn More
ACROBiosystems Recombinant Protein;Human Angiopoietin-2 / ANGPT2 (275-496) Protein;HEK293;100UG;AN2,ANGPT2,ANGPT-2,AGPT2,AGPT-2,ANG2,ANG-2,Angiopoietin-2,Angiopoietin 2
Supplier:  ACROBiosystems AN2H5242100UG
Encompass_Preferred
Recombinant Protein;Human Angiopoietin-2 / ANGPT2 (275-496) Protein;100UG;Human ANGPT2 (275-496), His Tag (AN2-H5242) is expressed from human 293 cells (HEK293). It contains AA Lys 275 - Phe 496 (Accession # AAI26201.1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 26.3 kDa. The protein migrates as 30-33 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-9060
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.