Learn More
ACROBiosystemsSupplier Diversity Partner Recombinant Protein;Human ALK-1 / ACVRL1 Protein;HEK293;100UG;AL1,ACVRL1,ACVRL-1,ACVRLK1,ALK-1,ALK1,HHT,HHT2,HHT-2,ORW2,ORW-2,SKR3,SKR-3,TSR-I,TSR-Ⅰ  DFS Item
Manufacturer:  ACROBiosystemsSupplier Diversity Partner AL1H5227100UG
Recombinant Protein;Human ALK-1 / ACVRL1 Protein;100UG;Human ALK-1, His Tag (AL1-H5227) is expressed from human 293 cells (HEK293). It contains AA Asp 22 - Gln 118 (Accession # NP_000011.2).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 12.3 kDa. The protein migrates as 26 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-9064