Learn More
ACROBiosystems Recombinant Protein;Human Akt1 Protein, His Tag & Strep Tag;HEK293;25UG;AK1,AKT1,PKB,RAC,RAC-PK-alpha,PKB alpha,AKT-1
Supplier:  ACROBiosystems AK1H528325UG
Encompass_Preferred
Recombinant Protein;Human Akt1 Protein, His Tag & Strep Tag;25UG;Human Akt1, His Tag, Strep Tag (AK1-H5283) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 480 (Accession # AAH00479).;This protein carries a polyhistidine tag at the C-terminus, followed by a twin strep tag. The protein has a calculated MW of 59.5 kDa. The protein migrates as 60-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-9067
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.