Learn More
ACROBiosystemsSupplier Diversity Partner Recombinant Protein;Biotinylated Human ErbB3 / Her3, ultra sensitivity (primary amine labeling);HEK293;25UG;ER3,ERBB3,ErbB-3,HER3,HER-3,LCCS2,MDA-BF-1,MGC88033,c-erbB-3,c-erbB3,erbB3-S,p180-ErbB3,p45-sErbB3,p85-sErbB3  DFS Item
Manufacturer:  ACROBiosystemsSupplier Diversity Partner ER3H822325UG
Recombinant Protein;Biotinylated Human ErbB3 / Her3, ultra sensitivity (primary amine labeling);25UG;MABSol Biotinylated Human ErbB3 / Her3 (ER3-H8223) is expressed from human HEK293 cells. It contains AA Ser 20 - Thr 643 (Accession # NP_001973.2). It is the biotinylated form of Human ErbB3 / Her3 Protein (Cat # ER3-H5223).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 71.5 kDa. The protein migrates as 100-110 kDa on a SDS-PAGE gel under reducing (R) condition due to glycosylation.
Catalog No. 50-165-9173