Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human EpCAM / TROP1, Fc Tag, ultra sensitivity (primary amine labeling);HEK293;25UG;EPM,EPCAM,DIAR5,DIAR-5,EGP-2,EGP2,EGP314,EGP-314,EGP40,EGP-40,ESA,GA733-2,HNPCC8,HNPCC-8,KS1/4,KSA,M4S1,MIC18,MIC-18,MK-1,MK1,TACSTD1,TACSTD-1,TROP1,TROP-1,CD326,CD-326
Supplier:  ACROBiosystems EPMH825425UG
Encompass_Preferred
Recombinant Protein;Biotinylated Human EpCAM / TROP1, Fc Tag, ultra sensitivity (primary amine labeling);25UG;MABSol Biotinylated Human EpCAM / TROP1, Fc Tag (EPM-H8254) is expressed from human HEK293 cells. It contains AA Gln 24 - Lys 265 (Accession # AAH14785.1). It is the biotinylated form of Human EpCAM protein, Fc Tag (Cat # EPM-H5254).;This protein carries a human IgG1 Fc fragment at the C-terminus. The protein has a calculated MW of 54 kDa. The protein migrates as 60-65 kDa on a SDS-PAGE gel under reducing (R) condition due to glycosylation.
Catalog No. 50-165-9178