Learn More
ACROBiosystems Recombinant Protein;ActiveMax Recombinant Human TGF-Beta 1 / TGFB1;HEK293;50UG;TG1,TGFB1,TGF B1,TGF-B1,TGF-beta 1,TGF beta1,TGFbeta1,CED,DPD1,DPD-1,LAP,TGFB,TGFbeta,TGF-1,TGF1,TGF-beta,TGF beta
Supplier:  ACROBiosystems TG1H421250UG
Encompass_Preferred
Recombinant Protein;ActiveMax Recombinant Human TGF-Beta 1 / TGFB1;50UG;Human TGFB1, Tag Free (TG1-H4212) is expressed from human 293 cells (HEK293). It contains AA Ala 279 - Ser 390 (Accession # P01137-1).;This protein carries no "tag". The protein has a calculated MW of 12.8 kDa (monomer). The protein migrates as 13 kDa (monomer) under reducing (R) condition (SDS-PAGE).;ActiveMax Human TGF-Beta 1 (Cat. No.TG1-H4212) inhibits the IL-4-dependent proliferation of TF-1 cells. The ED50 for this effect is 0.390-0.476 pg/ml (Routinely tested).
Catalog No. 50-165-9192