Learn More
ACROBiosystems ACROBiosystems Human Nectin-4 Protein, Fc Tag
Supplier: ACROBiosystems NE4H5255100UG
Recombinant Protein;100UG;Human Nectin-4, Fc Tag (NE4-H5255) is expressed from human 293 cells (HEK293). It contains AA Gly 32 - Ser 349 (Accession # Q96NY8-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 60.5 kDa. The protein migrates as 65-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Nectin-4, Fc Tag (Cat. No. NE4-H5255) at 5 μg/mL (100 μL/well) can bind Human Nectin-1, His Tag (Cat. No. PV1-H5223) with a linear range of 0.313-5 μg/mL (QC tested).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.