Learn More
ACROBiosystems ACROBiosystems Human BTLA (31-150) Protein, Mouse IgG2a Fc Tag
Supplier:  ACROBiosystems BTAH5258100UG
Encompass_Preferred
Recombinant Protein;100UG;Human BTLA (31-150), Mouse IgG2a Fc Tag (BTA-H5258) is expressed from human 293 cells (HEK293). It contains AA Lys 31 - Ser 150 (Accession # AAP44003.1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 40.6 kDa.
Catalog No. 50-195-6426