Learn More
ACROBiosystems ACROBiosystems ActiveMax Human Erythropoietin / EPO Protein, Tag Free
Supplier: ACROBiosystems EPOH421420UG
Recombinant Protein;20UG;ActiveMax Human EPO, Tag Free (EPO-H4214) is expressed from human 293 cells (HEK293). It contains AA Ala 28 - Arg 193 (Accession # AAH93628.1).;This protein carries no "tag". The protein has a calculated MW of 18.4 kDa. The protein migrates as 30-37 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.