Learn More
ACROBiosystemsSupplier Diversity Partner ACROBiosystems Human Flt-3 Ligand Protein, His Tag
Manufacturer:  ACROBiosystemsSupplier Diversity Partner FLLH522320UG
Encompass_Preferred
Recombinant Protein;20UG;Human Flt-3 Ligand, His Tag (FLL-H5223) is expressed from human 293 cells (HEK293). It contains AA Thr 27 - Pro 185 (Accession # P49771-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 19.9 kDa. The protein migrates as 28-30 kDa and 31-33 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Flt-3, Fc Tag (Cat. No. FL3-H5258) at 5 μg/mL (100 μL/well) can bind Human Flt-3 Ligand, His Tag (Cat. No. FLL-H5223) with a linear range of 0.1-0.6 ng/mL (QC tested).
Catalog No. 50-195-6489