Learn More
ACROBiosystems ACROBiosystems Biotinylated Cynomolgus PD-1 / PDCD1 Protein, His,Avitag™
Supplier: ACROBiosystems PD1C82E625UG
Recombinant Protein;25UG;Biotinylated Cynomolgus PD-1, His,Avitag (PD1-C82E6) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accession # B0LAJ3).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 19.5 kDa. The protein migrates as 30-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Cynomolgus PD-1, His,Avitag (Cat. No. PD1-C82E6) at 1 μg/mL (100 μL/well) on streptavidin precoated (0.5 μg/well) plate, can bind Cynomolgus / Rhesus macaque PD-L1, Fc Tag (Cat. No. PD1-C5253) with a linear range of 0.2-5 ng/mL (QC tested).;Immobilized Biotinylated Cynomolgus PD-1, His,Avitag (Cat. No. PD1-C82E6) at 1 μg/mL (100 μL/well) on streptavidin precoated (0.5 μg/well) plate, can bind Nivolumab with a linear range of 1-20 ng/mL (Routinely tested).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.