Learn More
ACROBiosystemsSupplier Diversity Partner ACROBiosystems Mouse PSMA / FOLH1 Protein, His Tag  DFS Item
Manufacturer:  ACROBiosystemsSupplier Diversity Partner PSAM5245100UG
Recombinant Protein;100UG;Mouse PSMA, His Tag (PSA-M5245) is expressed from human 293 cells (HEK293). It contains AA Ile 44 - Ala 752 (Accession # O35409-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 81.5 kDa. The protein migrates as 90-115 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6520