Learn More
ACROBiosystems ACROBiosystems Cynomolgus PSMA / FOLH1 Protein, His Tag
Supplier:  ACROBiosystems PSAC5247100UG
Encompass_Preferred
Recombinant Protein;100UG;Cynomolgus PSMA, His Tag (PSA-C5247) is expressed from human 293 cells (HEK293). It contains AA Lys 44 - Ala 750 (Accession # A0A2K5VNZ0-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 81.4 kDa.
Catalog No. 50-195-6523