Learn More
ACROBiosystemsSupplier Diversity Partner ACROBiosystems ActiveMax Human IL-7 Protein, Tag Free
Manufacturer:  ACROBiosystemsSupplier Diversity Partner IL7H421920UG
Encompass_Preferred
Recombinant Protein;20UG;ActiveMax Human IL-7, Tag Free (IL7-H4219) is expressed from human 293 cells (HEK293). It contains AA Asp 26 - His 177 (Accession # P13232).;This protein carries no "tag". The protein has a calculated MW of 17.4 kDa. The protein migrates as 24 kDa and 25-28 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.;Immobilized ActiveMax Human IL-7, Tag Free (Cat. No. IL7-H4219) at 2 μg/mL (100 μL/well) can bind Human IL-7 R alpha, Fc Tag (Cat. No. ILA-H5258) with a linear range of 1-16 ng/mL (QC tested).;Captured Human IL-7 R alpha, Fc Tag (Cat. No. ILA-H5258) on CM5 chip via anti-human IgG Fc antibodies surface can bind ActiveMax Human IL-7, Tag Free (Cat. No. IL7-H4219) with an affinity constant of 23.9 nM as determined in a SPR assay (Biacore T200) (Routinely tested).;Captured Human IL-7 R alpha, Mouse IgG2a Fc Tag (Cat. No. IL7-H5258) on CM5 chip via anti-mouse antibodies surface can bind ActiveMax Human IL-7, Tag Free (Cat. No. IL7-H4
Catalog No. 50-195-6545