Learn More
ACROBiosystemsSupplier Diversity Partner ACROBiosystems Human Siglec-10 Protein, Fc Tag  DFS Item
Manufacturer:  ACROBiosystemsSupplier Diversity Partner SI0H5253100UG
Recombinant Protein;100UG;Human Siglec-10, Fc Tag (SI0-H5253) is expressed from human 293 cells (HEK293). It contains AA Met 17 - Thr 546 (Accession # Q96LC7-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 84.6 kDa. The protein migrates as 90-115 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Siglec-10, Fc Tag (Cat. No. SI0-H5253) at 5 μg/mL (100 μL/well) can bind Anti-Human Siglec-10 MAb, human IgG with a linear range of 0.6-10 ng/mL (QC tested).
Catalog No. 50-195-6588