Learn More
ACROBiosystems ACROBiosystems Biotinylated Human IL-2 Protein, His,Avitag™
Manufacturer:  ACROBiosystems IL2H82E425UG
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human IL-2, His,Avitag (IL2-H82E4) is expressed from human 293 cells (HEK293). It contains AA Ala 21 - Thr 153 (Accession # P60568-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 19.1 kDa. The protein migrates as 19 kDa and 20 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-2 R alpha, Fc Tag (Cat. No. ILA-H5251) at 5 μg/mL (100 μL/well) can bind Biotinylated Human IL-2, His,Avitag (Cat. No. IL2-H82E4) with a linear range of 0.6-5 ng/mL (QC tested).
Catalog No. 50-195-6605