Learn More
ACROBiosystems ACROBiosystems Human HMGB1 Protein, His Tag
Supplier: ACROBiosystems HM1H5220100UG
Recombinant Protein;100UG;Human HMGB1, His Tag (HM1-H5220) is expressed from human 293 cells (HEK293). It contains AA Gly 2 - Glu 215 (Accession # NP_002119.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.0 kDa. The protein migrates as 32 kDa and 35 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.;Human HMGB1, His Tag (Cat. No. HM1-H5220) stimulates production of TNF-α in Raw-246.7 cells. The EC50 for this effect is 0.7855-0.8342 μg/mL (Routinely tested).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.