Learn More
ACROBiosystemsSupplier Diversity Partner ACROBiosystems Human IL-17E / IL-25 Protein, His Tag  DFS Item
Manufacturer:  ACROBiosystemsSupplier Diversity Partner IL5H422120UG
Recombinant Protein;20UG;Human IL-17E, His Tag (IL5-H4221) is expressed from human 293 cells (HEK293). It contains AA Tyr 33 - Gly 177 (Accession # NP_073626.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 17.6 kDa. The protein migrates as 23-28 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.
Catalog No. 50-195-6672